Assay Details
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033888.1 | NP_001029060.1 | 2 - 3 | 111 | 109 | |
XM_006228565.2 | XP_006228627.1 | 1 - 2 | 3275 | 109 | ||
XM_006228566.2 | XP_006228628.1 | 1 - 2 | 3275 | 109 | ||
XM_006228567.2 | XP_006228629.1 | 2 - 3 | 334 | 109 | ||
XM_006228568.2 | XP_006228630.1 | 2 - 3 | 540 | 109 | ||
GenBank mRNA | AB210107.1 | - | 2 - 3 | 111 | 109 | |
BC129073.1 | - | 2 - 3 | 221 | 7189989 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228568.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
