Assay Details
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033888.1 | NP_001029060.1 | 5 - 6 | 398 | 61 | |
XM_006228565.2 | XP_006228627.1 | 4 - 5 | 3562 | 61 | ||
XM_006228566.2 | XP_006228628.1 | 4 - 5 | 3562 | 61 | ||
XM_006228567.2 | XP_006228629.1 | 5 - 6 | 621 | 61 | ||
XM_006228568.2 | XP_006228630.1 | 5 - 6 | 827 | 61 | ||
GenBank mRNA | AB210107.1 | - | 5 - 6 | 398 | 61 | |
BC129073.1 | - | 5 - 6 | 508 | 7189989 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
