Assay Details
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014243.1 | NP_001014265.1 | 11 - 12 | 1282 | 65 | |
XM_006228450.2 | XP_006228512.1 | 11 - 12 | 2213 | 65 | ||
GenBank mRNA | BC087051.1 | - | 11 - 12 | 1282 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
