Assay Details
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014243.1 | NP_001014265.1 | 6 - 7 | 771 | 74 | |
XM_006228450.2 | XP_006228512.1 | 6 - 7 | 1702 | 74 | ||
GenBank mRNA | BC087051.1 | - | 6 - 7 | 771 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
