Assay Details
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228575.2 | XP_006228637.1 | 1 - 2 | 89 | 56 | |
GenBank mRNA | BC081858.1 | - | 1 - 2 | 63 | 56 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC081858.1, XM_006228575.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
