Assay Details
Gene Symbol: Grin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022797.1 | NP_073634.1 | 1 - 2 | 704 | 122 | |
GenBank mRNA | L31612.1 | - | 1 - 2 | 704 | 122 | |
U08260.1 | - | 1 - 2 | 50 | 122 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the L31612.1, NM_022797.1, U08260.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
