Assay Details
Gene Symbol: Tph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100634.2 | NP_001094104.1 | 3 - 4 | 239 | 145 | |
XM_006229218.2 | XP_006229280.1 | 2 - 3 | 365 | 145 | ||
XM_006229219.1 | XP_006229281.1 | 3 - 4 | 278 | 145 | ||
GenBank mRNA | CB765437.1 | - | 3 - 4 | 193 | 145 | |
X53501.1 | - | 3 - 4 | 239 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
