Assay Details
Gene Symbol: Tph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100634.2 | NP_001094104.1 | 4 - 5 | 420 | 147 | |
XM_006229218.2 | XP_006229280.1 | 3 - 4 | 546 | 147 | ||
XM_006229219.1 | XP_006229281.1 | 4 - 5 | 459 | 147 | ||
GenBank mRNA | X53501.1 | - | 4 - 5 | 420 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
