Assay Details
Gene Symbol: Tph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100634.2 | NP_001094104.1 | 8 - 9 | 922 | 66 | |
XM_006229218.2 | XP_006229280.1 | 7 - 8 | 1048 | 66 | ||
XM_006229219.1 | XP_006229281.1 | 8 - 9 | 961 | 66 | ||
GenBank mRNA | CB609064.1 | - | 1 - 2 | 37 | 66 | |
CB796568.1 | - | 3 - 4 | 384 | 66 | ||
X53501.1 | - | 8 - 9 | 922 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
