Assay Details
Gene Symbol: Mamstr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001076631.5 | XP_001076631.2 | 9 - 10 | 1951 | 97 | |
XM_006229210.2 | XP_006229272.1 | 9 - 10 | 2336 | 97 | ||
XM_006229211.2 | XP_006229273.1 | 9 - 10 | 1895 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
