Assay Details
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228857.2 | XP_006228919.1 | 10 - 11 | 1584 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
