Assay Details
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107512.1 | NP_001100982.1 | 11 - 12 | 1342 | 64 | |
XM_006229069.1 | XP_006229131.1 | 11 - 12 | 1297 | 64 | ||
XM_006229070.2 | XP_006229132.1 | 11 - 12 | 1299 | 64 | ||
XM_008759378.1 | XP_008757600.1 | 11 - 12 | 1381 | 64 | ||
GenBank mRNA | BC168947.1 | - | 11 - 12 | 1291 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
