Assay Details
Gene Symbol: Adamts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024400.2 | NP_077376.2 | 3 - 4 | 1589 | 68 | |
GenBank mRNA | AF149118.1 | - | 3 - 4 | 1602 | 68 | |
AF304446.1 | - | 3 - 4 | 1160 | 68 | ||
BC080237.1 | - | 3 - 4 | 1589 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
