Assay Details
Gene Symbol: Sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017050.1 | NP_058746.1 | 2 - 3 | 258 | 97 | |
GenBank mRNA | BC082800.1 | - | 2 - 3 | 238 | 97 | |
FQ209495.1 | - | 2 - 3 | 259 | 97 | ||
FQ209941.1 | - | 2 - 3 | 254 | 97 | ||
M21060.1 | - | 2 - 3 | 161 | 97 | ||
M25157.1 | - | 2 - 3 | 97 | |||
X05634.1 | - | 2 - 3 | 157 | 97 | ||
Y00404.1 | - | 2 - 3 | 258 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
