Assay Details
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108490.1 | NP_001101960.1 | 10 - 11 | 1053 | 91 | |
XM_006229441.2 | XP_006229503.1 | 9 - 10 | 935 | 91 | ||
XM_008759543.1 | XP_008757765.1 | 8 - 9 | 1315 | 91 | ||
XM_008759544.1 | XP_008757766.1 | 9 - 10 | 907 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P417031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31971 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P417031 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P417031 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P417031 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P417031 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49858 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92632 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37643 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10708336 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708332 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708335 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708333 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708337 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708334 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708330 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708331 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708329 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10708327 | - | - |
Rat Genome 230 2.0 Array | 1374151_at | - | - |
Rat Genome 230 2.0 Array | 1379482_at | - | - |
Rat Genome 230 2.0 Array | 1394596_at | - | - |
Rat Genome 230 2.0 Array | 1374151_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394596_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379482_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379482_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379482_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100014923.rn.1 | TC0100001892.rn.1 | RTA TC |
Rat Expression Set 230 | 1379482_at | - | - |
Rat Expression Set 230 | 1379482_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708327 | - | - |
Rat Genome U34 Set | rc_AI233614_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22163154 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371959 | - | - |
