Assay Details
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108915.1 | NP_001102385.1 | 3 - 4 | 663 | 73 | |
XM_006229350.2 | XP_006229412.1 | 4 - 5 | 831 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108915.1, XM_006229350.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
