Assay Details
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108915.1 | NP_001102385.1 | 4 - 5 | 927 | 124 | |
XM_006229349.2 | XP_006229411.1 | 5 - 6 | 1129 | 124 | ||
XM_006229350.2 | XP_006229412.1 | 5 - 6 | 1095 | 124 | ||
XM_008759545.1 | XP_008757767.1 | 4 - 5 | 2073 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
