Assay Details
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017181.2 | NP_058877.1 | 7 - 8 | 702 | 59 | |
GenBank mRNA | BC076381.1 | - | 7 - 8 | 702 | 59 | |
FQ219602.1 | - | 7 - 8 | 660 | 59 | ||
M77694.1 | - | 7 - 8 | 624 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
