Assay Details
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031238.2 | NP_112517.1 | 8 - 9 | 1192 | 135 | |
XM_006229480.2 | XP_006229542.1 | 8 - 9 | 980 | 135 | ||
XM_006229481.2 | XP_006229543.1 | 8 - 9 | 1073 | 135 | ||
XM_006229483.2 | XP_006229545.1 | 8 - 9 | 1179 | 135 | ||
XM_006229485.1 | XP_006229547.1 | 6 - 7 | 723 | 135 | ||
XM_006229486.2 | XP_006229548.1 | 6 - 7 | 647 | 135 | ||
GenBank mRNA | AB008159.2 | - | 5 - 6 | 637 | 135 | |
AF009604.1 | - | 6 - 7 | 676 | 135 | ||
AF227439.1 | - | 8 - 9 | 1195 | 135 | ||
BC078800.1 | - | 8 - 9 | 1195 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P272894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55268 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P272894 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P272894 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P272894 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P272894 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702152136 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19839 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14490 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46457 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29874 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31485 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104157 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93904 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20645 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702152136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10708339 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708347 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708342 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708341 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708346 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708345 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708348 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708344 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10708338 | - | - |
Rat Genome 230 2.0 Array | 1368931_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368931_at | - | - |
Affymetrix HT Rat Focus Array | 1368931_at | - | - |
Affymetrix HT Rat Focus Array | 1368931_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100014960.rn.1 | TC0100001896.rn.1 | RTA TC |
Rat Expression Set 230 | 1368931_at | - | - |
Rat Expression Set 230 | 1368931_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708338 | - | - |
Rat Genome U34 Set | AF009604_at | - | - |
Rat Genome U34 Set | rc_AA943234_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21528058 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365281 | - | - |
RatRef-12 Expression BeadChip | ILMN_51568 | - | - |
