Assay Details
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177481.1 | NP_803434.1 | 8 - 9 | 1681 | 66 | |
XM_006229378.2 | XP_006229440.1 | 8 - 9 | 1846 | 66 | ||
GenBank mRNA | AF239219.1 | - | 8 - 9 | 1681 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
