Assay Details
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177481.1 | NP_803434.1 | 9 - 10 | 1748 | 73 | |
XM_006229378.2 | XP_006229440.1 | 9 - 10 | 1913 | 73 | ||
GenBank mRNA | AF239219.1 | - | 9 - 10 | 1748 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P496255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P496255 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P496255 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P496255 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P496255 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50020 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51416 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20852 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88177 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65413 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37181 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32052 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102861 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61950 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34323 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10722825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722823 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10722818 | - | - |
Rat Genome 230 2.0 Array | 1373734_at | - | - |
Rat Genome 230 2.0 Array | 1370571_at | - | - |
Rat Genome 230 2.0 Array | 1373734_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370571_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370571_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373734_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373734_at | - | - |
Affymetrix HT Rat Focus Array | 1370571_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100046201.rn.1 | TC0100005786.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100046203.rn.1 | TC0100005786.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100046202.rn.1 | TC0100005786.rn.1 | RTA TC |
Rat Expression Set 230 | 1373734_at | - | - |
Rat Expression Set 230 | 1370571_PM_at | - | - |
Rat Expression Set 230 | 1373734_PM_at | - | - |
Rat Expression Set 230 | 1370571_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10722818 | - | - |
Rat Genome U34 Set | rc_AI101171_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21680302 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362594 | - | - |
RatRef-12 Expression BeadChip | ILMN_61669 | - | - |
