Assay Details
Gene Symbol: Idh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014161.1 | NP_001014183.1 | 8 - 9 | 1108 | 67 | |
GenBank mRNA | BC076398.1 | - | 8 - 9 | 1108 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
