Assay Details
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007731.1 | NP_001007732.1 | 2 - 3 | 473 | 169 | |
GenBank mRNA | BC078959.1 | - | 2 - 3 | 473 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
