Assay Details
Gene Symbol: Gnaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031036.1 | NP_112298.1 | 3 - 4 | 466 | 75 | |
GenBank mRNA | AF234260.1 | - | 3 - 4 | 466 | 75 | |
DQ120477.1 | - | 3 - 4 | 431 | 75 | ||
DQ120478.1 | - | 3 - 4 | 431 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
