Assay Details
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127542.1 | NP_001121014.1 | 4 - 5 | 529 | 72 | |
GenBank mRNA | BC161922.1 | - | 4 - 5 | 515 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
