Assay Details
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134334.2 | NP_599161.2 | 7 - 8 | 1063 | 79 | |
GenBank mRNA | BC062032.1 | - | 7 - 8 | 1063 | 79 | |
FQ213891.1 | - | 2 - 3 | 189 | 79 | ||
FQ228180.1 | - | 3 - 4 | 402 | 79 | ||
FQ232703.1 | - | 3 - 4 | 402 | 79 | ||
X54467.1 | - | 7 - 8 | 1008 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
