Assay Details
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134334.2 | NP_599161.2 | 5 - 6 | 801 | 82 | |
GenBank mRNA | BC062032.1 | - | 5 - 6 | 801 | 82 | |
FQ228180.1 | - | 1 - 2 | 140 | 82 | ||
FQ232703.1 | - | 1 - 2 | 140 | 82 | ||
X54467.1 | - | 5 - 6 | 746 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
