Assay Details
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230922.2 | XP_006230984.1 | 2 - 3 | 344 | 151 | |
XM_006230923.2 | XP_006230985.1 | 2 - 3 | 604 | 151 | ||
XM_006230924.2 | XP_006230986.1 | 2 - 3 | 401 | 151 | ||
XM_006230925.2 | XP_006230987.1 | 2 - 3 | 368 | 151 | ||
XM_008760190.1 | XP_008758412.1 | 2 - 3 | 247 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
