Assay Details
Gene Symbol: Pc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012744.2 | NP_036876.2 | 19 - 20 | 3352 | 72 | |
XM_006230689.2 | XP_006230751.1 | 20 - 21 | 3583 | 72 | ||
XM_006230690.2 | XP_006230752.1 | 19 - 20 | 3698 | 72 | ||
XM_006230691.2 | XP_006230753.1 | 20 - 21 | 3405 | 72 | ||
XM_006230692.2 | XP_006230754.1 | 19 - 20 | 3375 | 72 | ||
XM_006230693.2 | XP_006230755.1 | 20 - 21 | 3507 | 72 | ||
XM_006230694.2 | XP_006230756.1 | 19 - 20 | 3374 | 72 | ||
XM_008760087.1 | XP_008758309.1 | 19 - 20 | 3480 | 72 | ||
GenBank mRNA | BC085680.1 | - | 19 - 20 | 3329 | 72 | |
U32314.1 | - | 19 - 20 | 3320 | 72 | ||
U36585.1 | - | 19 - 20 | 3352 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
pyruvate metabolic process
gluconeogenesis
oxaloacetate metabolic process
lipid metabolic process
negative regulation of gene expression
viral RNA genome packaging
positive regulation by host of viral release from host cell
positive regulation by host of viral process
DNA binding
biotin carboxylase activity
pyruvate carboxylase activity
ATP binding
biotin binding
carboxylic acid binding
metal ion binding
pyruvate metabolic process
gluconeogenesis
oxaloacetate metabolic process
lipid metabolic process
negative regulation of gene expression
viral RNA genome packaging
positive regulation by host of viral release from host cell
positive regulation by host of viral process
DNA binding
biotin carboxylase activity
pyruvate carboxylase activity
ATP binding
biotin binding
carboxylic acid binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173274 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085641 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085641 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085641 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P533312 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700329916 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701882545 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701878528 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90928 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100850 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30906 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91221 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98769 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34206 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45350 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701882545 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701878528 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700329916 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10712886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712874 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712875 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712871 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712872 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712868 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10712867 | - | - |
Rat Genome 230 2.0 Array | 1386917_at | - | - |
Rat Genome 230 2.0 Array | 1386917_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386917_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386917_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100024344.rn.1 | TC0100003007.rn.1 | RTA TC |
Rat Expression Set 230 | 1386917_PM_at | - | - |
Rat Expression Set 230 | 1386917_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712867 | - | - |
Rat Genome U34 Set | U32314_at | - | - |
Rat Genome U34 Set | U32314_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21567976 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368486 | - | - |
RatRef-12 Expression BeadChip | ILMN_58548 | - | - |
