Assay Details
Gene Symbol: Mtg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749002.3 | XP_003749050.1 | 1 - 2 | 164 | 148 | |
XM_006230584.2 | XP_006230646.1 | 1 - 2 | 165 | 148 | ||
GenBank mRNA | FQ212842.1 | - | 1 - 2 | 130 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
