Assay Details
Gene Symbol: Nudt22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199090.2 | NP_954521.1 | 4 - 5 | 791 | 84 | |
XM_006230760.2 | XP_006230822.1 | 3 - 4 | 693 | 84 | ||
GenBank mRNA | BC060593.1 | - | 4 - 5 | 792 | 84 | |
FQ212766.1 | - | 4 - 5 | 738 | 84 | ||
FQ215165.1 | - | 4 - 5 | 719 | 84 | ||
FQ219356.1 | - | 4 - 5 | 742 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
