Assay Details
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013032.3 | NP_037164.3 | 9 - 10 | 1085 | 86 | |
GenBank mRNA | AF038571.1 | - | 2 - 3 | 198 | 86 | |
BC061743.1 | - | 9 - 10 | 1085 | 86 | ||
D63772.1 | - | 1116 | 86 | |||
L35558.1 | - | 4 - 5 | 531 | 86 | ||
U21104.1 | - | 9 - 10 | 1097 | 86 | ||
U39555.1 | - | 9 - 10 | 1175 | 86 | ||
X94255.1 | - | 9 - 10 | 1142 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
