Assay Details
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106375.2 | NP_001099845.2 | 10 - 11 | 1509 | 109 | |
XM_006231274.2 | XP_006231336.1 | 11 - 12 | 1669 | 109 | ||
GenBank mRNA | BC169061.1 | - | 10 - 11 | 1509 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
