Assay Details
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106375.2 | NP_001099845.2 | 11 - 12 | 1741 | 104 | |
XM_006231274.2 | XP_006231336.1 | 12 - 13 | 1901 | 104 | ||
GenBank mRNA | BC169061.1 | - | 11 - 12 | 1741 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P038852 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P513050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20699 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P513050 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038852 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P038852 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P513050 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P038852 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P513050 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119160 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1071228 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40922 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10714823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714828 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10714818 | - | - |
Rat Genome 230 2.0 Array | 1376047_at | - | - |
Rat Genome 230 2.0 Array | 1395721_at | - | - |
Rat Genome 230 2.0 Array | 1395721_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376047_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376047_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376047_at | - | - |
Affymetrix HT Rat Focus Array | 1395721_at | - | - |
Affymetrix HT Rat Focus Array | 1395721_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100028316.rn.1 | TC0100003486.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100028315.rn.1 | TC0100003486.rn.1 | RTA TC |
Rat Expression Set 230 | 1376047_PM_at | - | - |
Rat Expression Set 230 | 1376047_at | - | - |
Rat Expression Set 230 | 1395721_PM_at | - | - |
Rat Expression Set 230 | 1395721_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714818 | - | - |
Rat Genome U34 Set | rc_AA893244_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21413907 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358624 | - | - |
