Assay Details
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760316.1 | XP_008758538.1 | 9 - 10 | 1965 | 81 | |
GenBank mRNA | BC079474.1 | - | 3 - 4 | 319 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
