Assay Details
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212520.1 | NP_997685.1 | 4 - 5 | 533 | 95 | |
XM_006228420.2 | XP_006228482.1 | 5 - 6 | 641 | 95 | ||
GenBank mRNA | AB162855.1 | - | 4 - 5 | 533 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
