Assay Details
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008775.1 | NP_001008775.1 | 8 - 9 | 613 | 90 | |
XM_006228776.2 | XP_006228838.1 | 7 - 8 | 630 | 90 | ||
XM_006228777.1 | XP_006228839.1 | 6 - 7 | 514 | 90 | ||
XM_006228779.2 | XP_006228841.1 | 6 - 7 | 531 | 90 | ||
GenBank mRNA | AY325237.1 | - | 675 | 90 | ||
AY325238.1 | - | 8 - 9 | 613 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
