Assay Details
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 6 - 7 | 522 | 87 | |
GenBank mRNA | BC063156.1 | - | 6 - 7 | 581 | 87 | |
X89968.1 | - | 6 - 7 | 522 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
