Assay Details
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751193.3 | XP_003751241.1 | 8 - 9 | 2404 | 106 | |
XM_006249412.2 | XP_006249474.1 | 8 - 9 | 2416 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
