Assay Details
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008292.1 | NP_001008293.1 | 3 - 4 | 335 | 79 | |
GenBank mRNA | BC086366.1 | - | 3 - 4 | 335 | 79 | |
FQ219826.1 | - | 3 - 4 | 328 | 79 |
Gene Symbol: LOC100360940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729540.3 | XP_002729586.2 | 2 | 365 | 79 | |
XM_008764180.1 | XP_008762402.1 | 1 | 152 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
