Assay Details
Gene Symbol: Gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201991.1 | NP_973720.1 | 2 - 3 | 208 | 84 | |
GenBank mRNA | AY513752.1 | - | 2 - 3 | 208 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
