Assay Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031151.2 | NP_112413.2 | 4 - 5 | 532 | 87 | |
GenBank mRNA | BC063165.1 | - | 4 - 5 | 532 | 87 | |
DQ402951.1 | - | 3 - 4 | 366 | 87 | ||
FQ213494.1 | - | 4 - 5 | 485 | 87 | ||
FQ213817.1 | - | 4 - 5 | 447 | 87 | ||
FQ215385.1 | - | 4 - 5 | 492 | 87 | ||
FQ215647.1 | - | 4 - 5 | 493 | 87 | ||
FQ216782.1 | - | 4 - 5 | 523 | 87 | ||
FQ217232.1 | - | 4 - 5 | 484 | 87 | ||
FQ224847.1 | - | 4 - 5 | 429 | 87 | ||
X04240.1 | - | 4 - 5 | 473 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
