Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107153.1 | NP_001100623.1 | 10 - 11 | 1075 | 72 | |
XM_008769337.1 | XP_008767559.1 | 10 - 11 | 1080 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
