Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107153.1 | NP_001100623.1 | 9 - 10 | 992 | 70 | |
XM_008769337.1 | XP_008767559.1 | 9 - 10 | 997 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
