Assay Details
Gene Symbol: Ncf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053734.2 | NP_446186.2 | 4 - 5 | 405 | 65 | |
GenBank mRNA | AF260779.1 | - | 3 - 4 | 333 | 65 | |
AF547392.1 | - | 4 - 5 | 405 | 65 | ||
AF547393.1 | - | 4 - 5 | 405 | 65 | ||
AY029167.1 | - | 4 - 5 | 405 | 65 | ||
BC061810.1 | - | 4 - 5 | 405 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
