Assay Details
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053971.3 | NP_446423.2 | 1 - 2 | 259 | 142 | |
XM_006249377.1 | XP_006249439.1 | 2 - 3 | 278 | 142 | ||
XM_006249378.2 | XP_006249440.1 | 2 - 3 | 312 | 142 | ||
GenBank mRNA | BC061784.1 | - | 1 - 2 | 255 | 142 | |
BC078761.1 | - | 1 - 2 | 262 | 142 | ||
BC097295.1 | - | 1 - 2 | 263 | 142 | ||
FQ229484.1 | - | 2 - 3 | 277 | 142 | ||
X87107.1 | - | 2 - 3 | 273 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
