Assay Details
Gene Symbol: Sumf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025125.2 | NP_001020296.2 | 2 - 3 | 253 | 160 | |
GenBank mRNA | BC091176.1 | - | 2 - 3 | 259 | 160 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
