Assay Details
Gene Symbol: Vsig10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271336.1 | NP_001258265.1 | 4 - 5 | 747 | 76 | |
XM_006249437.2 | XP_006249499.1 | 3 - 4 | 3071 | 76 | ||
XM_006249438.2 | XP_006249500.1 | 3 - 4 | 3071 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
