Assay Details
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053742.1 | NP_446194.1 | 6 - 7 | 398 | 66 | |
XM_006249522.2 | XP_006249584.1 | 6 - 7 | 442 | 66 | ||
XM_006249523.2 | XP_006249585.1 | 6 - 7 | 442 | 66 | ||
GenBank mRNA | BC061538.1 | - | 6 - 7 | 430 | 66 | |
D21132.1 | - | 6 - 7 | 398 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
