Assay Details
Gene Symbol: Cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033684.1 | NP_001028856.1 | 13 - 14 | 1537 | 88 | |
GenBank mRNA | BC091282.1 | - | 7 - 8 | 659 | 88 | |
BC104703.1 | - | 13 - 14 | 1537 | 88 | ||
FQ216833.1 | - | 13 - 14 | 1555 | 88 | ||
FQ218432.1 | - | 13 - 14 | 1584 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
